Learn More
[2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, LoGiCal
Product Code. 30462856
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462856 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462856 Supplier LoGiCal Supplier No. LGCFOR0006.04
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2-[(2,6-Dichlorophenyl)amino]phenyl)methanol
CAS 27204-57-5
Analyte or Component Names (2-[(2,6-Dichlorophenyl)amino]phenyl)methanol
Shipping Condition Room Temperature
Molecular Formula C13 H11 Cl2 N O
Certifications/Compliance ISO 9001
Synonym [2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, Diclofenac Sodium Imp. C (EP), Diclofenac Imp. C (EP), [2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, Diclofenac Sodium Impurity C, Diclofenac Impurity C
InChI Formula InChI=1S/C13H11Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-7,16-17H,8H2
Solution Type Neat
SMILES OCc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name [2-(2,6-dichloroanilino)phenyl]methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 268.14
Quantity 10 mg
Formula Weight 267.0218 g/mol