Learn More
2-(2,4,5-Trifluorophenyl)ethanol, 97%, Thermo Scientific™
Product Code. 15455437
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455437 250 mg 250mg
15445437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15455437 Supplier Thermo Scientific Alfa Aesar Supplier No. H32686.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 883267-70-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD06660356
InChI Key KHFXXIXHSGJQOW-UHFFFAOYSA-N
PubChem CID 17750839
IUPAC Name 2-(2,4,5-trifluorophenyl)ethanol
SMILES C1=C(C(=CC(=C1F)F)F)CCO
Specifications
Refractive Index 1.4732
Quantity 250 mg
Synonym 2-2,4,5-trifluorophenyl ethanol, 2-2,4,5-trifluorophenyl-ethanol, 2,4,5-trifluorobenzeneethanol, 2-2,4,5-trifluorophenyl ethan-1-ol, 2,4,5-trifluorophenylethanol, 2,4,5-trifluorophenethyl alcohol, benzeneethanol, 2,4,5-trifluoro, benzeneethanol,2,4,5-trifluoro, 2-2,4,5-tris fluoranyl phenyl ethanol
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-(2,4,5-Trifluorophenyl)ethanol