accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 883267-70-7

IUPAC Name: 2-(2,4,5-trifluorophenyl)ethan-1-ol
Synonyms: 2-(2,4,5-trifluorophenyl)ethanol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: KHFXXIXHSGJQOW-UHFFFAOYSA-N
SMILES: OCCC1=CC(F)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results