Learn More
2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene
Specifications
Specifications
| Chemical Name or Material | 2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene |
| CAS | 141214-18-8 |
| Analyte or Component Names | 2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H24 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propofol Imp. G (EP), Propofol USP Related Compound C, Propofol USP RC C, 2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene, Propofol Related Compound C, Propofol Impurity G |
| InChI Formula | InChI=1S/C15H24O/c1-10(2)13-8-7-9-14(11(3)4)15(13)16-12(5)6/h7-12H,1-6H3 |
| SMILES | CC(C)Oc1c(cccc1C(C)C)C(C)C |
| IUPAC Name | 1,3-di(propan-2-yl)-2-propan-2-yloxybenzene |
| Show More |