missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2696553
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10772283

Brand: Thermo Scientific Maybridge CC09101CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3529-57-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD05664666
InChI Key ZGDGZMOKXTUMEV-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
PubChem CID 2795202
IUPAC Name 2,1,3-benzothiadiazole-4-carboxylic acid
SMILES C1=CC2=NSN=C2C(=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.