accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NSNOOH

CAS RN 3529-57-5

IUPAC Name: 2,1,3-benzothiadiazole-4-carboxylic acid
Synonyms: 2,1,3-benzothiadiazole-4-carboxylic acid
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: ZGDGZMOKXTUMEV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=NSN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results