Learn More
(1S,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline, Mikromol™
Description
(1S,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline
Specifications
Specifications
| Chemical Name or Material | (1S,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline |
| CAS | 2575516-78-6 |
| Analyte or Component Names | (1S,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1S,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline |
| InChI Formula | InChI=1S/C16H17NO3/c1-17-9-15(20)14-8-12(19)5-6-13(14)16(17)10-3-2-4-11(18)7-10/h2-8,15-16,18-20H,9H2,1H3/t15-,16-/m0/s1 |
| SMILES | CN1C[C@H](O)c2cc(O)ccc2[C@@H]1c3cccc(O)c3 |
| IUPAC Name | (1S,4R)-1-(3-hydroxyphenyl)-2-methyl-3,4-dihydro-1H-isoquinoline-4,6-diol |
| Show More |