Learn More
(1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,N-dimethylpentan-1-amine Hydrochloride, Mikromol™
Description
(1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,N-dimethylpentan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (1RS)-1-(1-(4-Chlorophenyl)cyclobutyl)-N,N-dimethylpentan-1-amine Hydrochloride |
| CAS | 2727060-94-6 |
| Analyte or Component Names | (1RS)-1-(1-(4-Chlorophenyl)cyclobutyl)-N,N-dimethylpentan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 Cl N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,N-dimethylpentan-1-amine Hydrochloride, Sibutramine Hydrochloride Monohydrate Imp. C (Pharmeuropa) as Hydrochloride, Sibutramine Related Compound C as Hydrochloride, Sibutramine Hydrochloride Monohydrate Impurity C, Sibutramine Impurity C |
| InChI Formula | InChI=1S/C17H26ClN.ClH/c1-4-5-7-16(19(2)3)17(12-6-13-17)14-8-10-15(18)11-9-14;/h8-11,16H,4-7,12-13H2,1-3H3;1H |
| SMILES | Cl.CCCCC(N(C)C)C1(CCC1)c2ccc(Cl)cc2 |
| IUPAC Name | 1-[1-(4-chlorophenyl)cyclobutyl]-N,N-dimethylpentan-1-amine;hydrochloride |
| Show More |