Learn More
(1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol (Tropine), Mikromol™
Product Code. 30050767
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050767 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050767 Supplier Mikromol Supplier No. MM0763.010025
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol (Tropine)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tropine
CAS 120-29-6
Analyte or Component Names Tropine
Shipping Condition Ice pack (-20°C)
Molecular Formula C8 H15 N O
Certifications/Compliance ISO 9001
Synonym Tropisetron Hydrochloride Imp. A (EP), Tropisetron Imp. A (EP), (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol, Tropine, Tropisetron Hydrochloride Impurity A, Tropisetron Impurity A
InChI Formula InChI=1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8+
SMILES CN1[C@@H]2CC[C@H]1C[C@@H](O)C2
IUPAC Name (1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular Weight (g/mol) 141.21
Quantity 25 mg
Formula Weight 141.1154 g/mol