Learn More
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] Chloride, Mikromol™
Description
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] Chloride
Specifications
Specifications
| Chemical Name or Material | Azoniaspironortropanol chloride |
| CAS | 3464-71-9 |
| Analyte or Component Names | Azoniaspironortropanol chloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C11 H20 N O . Cl |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trospium Chloride Imp. C (EP), Trospium Chloride USP Related Compound C, Trospium Chloride USP RC C, 3-hydroxy-Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride, 3alpha-Hydroxynortropane-8-spiro-1'-pyrrolidinium chloride, (1alpha,3beta,5alpha)-3-Hydroxy-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride (1:1), (1R,3r,5S)-3-hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], Trospium Imp. C, Trospium RC C (USP), Azoniaspironortropanol chloride, Trospium Related Compound C as Chloride, Trospium Chloride Impurity C as Chloride, Trospium Impurity C as Chloride |
| InChI Formula | InChI=1S/C11H20NO.ClH/c13-11-7-9-3-4-10(8-11)12(9)5-1-2-6-12;/h9-11,13H,1-8H2;1H/q+1;/p-1/t9-,10+,11+; |
| SMILES | [Cl-].O[C@H]1C[C@H]2CC[C@@H](C1)[N+]23CCCC3 |
| IUPAC Name | (1S,5R)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-ol;chloride |
| Show More |