missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3R)-N-(4-Methoxybenzyl)-(4S)-amino-1,2,3-trihydroxy-(5R)-iodocyclopentane 3,4-Carbamate 1,2-Cyclohexyl Ketal, TRC

Product Code. 30474411
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474411 5 mg 5mg
30474048 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474411 Supplier Toronto Research Chemicals Supplier No. TRCM2261155MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,2R,3R)-N-(4-Methoxybenzyl)-(4S)-amino-1,2,3-trihydroxy-(5R)-iodocyclopentane 3,4-Carbamate 1,2-Cyclohexyl Ketal
Analyte or Component Names (1R,2R,3R)-N-(4-Methoxybenzyl)-(4S)-amino-1,2,3-trihydroxy-(5R)-iodocyclopentane 3,4-Carbamate 1,2-Cyclohexyl Ketal
Shipping Condition Room Temperature
Molecular Formula C20 H24 I N O5
Synonym (1R,2R,3R,4S,5R)-Hexahydro-7'-iodo-6'-[(4-methoxyphenyl)methyl]-spiro[cyclohexane-1,2'-[5H-1,3]dioxolo[3,4]cyclopent[1,2-d]oxazol]-5'-one
InChI Formula InChI=1S/C20H24INO5/c1-24-13-7-5-12(6-8-13)11-22-15-14(21)16-18(17(15)25-19(22)23)27-20(26-16)9-3-2-4-10-20/h5-8,14-18H,2-4,9-11H2,1H3/t14-,15-,16+,17-,18+/m1/s1
Solution Type Neat
SMILES COc1ccc(CN2[C@@H]3[C@@H](I)[C@@H]4OC5(CCCCC5)O[C@@H]4[C@@H]3OC2=O)cc1
IUPAC Name (1R,2R,6R,7R,8S)-7-iodo-9-[(4-methoxyphenyl)methyl]spiro[3,5,11-trioxa-9-azatricyclo[6.3.0.02,6]undecane-4,1'-cyclohexane]-10-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 485.313
Quantity 5 mg
Formula Weight 485.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.