Learn More
(1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, Mikromol™
Product Code. 30458269
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458269 4 x 25 mg -
17971386 100 mg 100mg
17971476 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30458269 Supplier Mikromol Supplier No. MM1342.0700254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
CAS 180272-45-1
Analyte or Component Names (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Shipping Condition Room Temperature
Molecular Formula C15 H15 N
Certifications/Compliance ISO 17025
Synonym Isoquinoline, 1,2,3,4-tetrahydro-1-phenyl-, (1R)-, Isoquinoline, 1,2,3,4-tetrahydro-1-phenyl-, (R)-, (-)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-Phenyl-1,2,3,4-tetrahydroisoquinoline, (R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
InChI Formula InChI=1S/C15H15N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-9,15-16H,10-11H2/t15-/m1/s1
Solution Type Neat
SMILES C1Cc2ccccc2[C@H](N1)c3ccccc3
IUPAC Name (1R)-1-phenyl-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol) 209.29
Quantity 4 x 25 mg
Formula Weight 209.1204 g/mol