missing translation for 'onlineSavingsMsg'
Learn More

D,L-Mevalonic Acid-d3 Dicyclohexylamine Salt, TRC

Product Code. 17918509
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17918509 1 mg 1mg
30133458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17918509 Supplier Toronto Research Chemicals Supplier No. TRCM3390221MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329796-98-6
Molecular Formula C18H32D3NO4
Molecular Weight (g/mol) 332.49
Synonym 3,5-Dihydroxy-3-(methyl-d3) Pentanoic Acid Dicyclohexylamine Salt, Hiochic Acid-d3 Dicyclohexylamine Salt, 3,5-Dihydroxy-3-(methyl-d3)valeric Acid Dicyclohexylamine Salt, 2,4-Dideoxy-3-C-(methyl-d3)
IUPAC Name dicyclohexylamine 3,5-dihydroxy-3-(methyl-d3)pentanoate
SMILES OCCC(O)(C([2H])([2H])[2H])CC(O)=O.C1(CCCCC1)NC2CCCCC2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H23N.C6H12O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(10,2-3-7)4-5(8)9/h11-13H,1-10H2;7,10H,2-4H2,1H3,(H,8,9)/i;1D3
Formula Weight 332.28
Chemical Name or Material D,L-Mevalonic Acid-d3 Dicyclohexylamine Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.