missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluorooctylamine, 97%, Thermo Scientific™

Product Code. 11383786
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383786 5 g 5g
11373786 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11383786 Supplier Thermo Scientific Alfa Aesar Supplier No. L16845.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H-Perfluorooctylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-29-9
Molecular Formula C8H4F15N
Molecular Weight (g/mol) 399.103
MDL Number MFCD00042460
InChI Key HZCZXRSVKNFILZ-UHFFFAOYSA-N
PubChem CID 136148
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)N

Specifications

Density 1.714
Boiling Point 149°C to 150°C
Flash Point None
Odor Amine-like
Linear Formula CF3(CF2)6CH2NH2
Refractive Index 1.305
Quantity 5 g
UN Number UN2735
Beilstein 1806966
Synonym 1h,1h-perfluorooctylamine, 1h,1h-pentadecafluorooctylamine, 1-octanamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluorooctylamine, 1h,1h-pentadecafluorooctylamin, 1h,1h-perfluorooctylamine 1g, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octylamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctylamine, 1-octanamine,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 399.1
Percent Purity 97%
Chemical Name or Material 1H,1H-Perfluorooctylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.