accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NFFFFFFFFFFFFFFF

CAS RN 307-29-9

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine
Molecular Weight (g/mol): 399.1
Molecular Formula: C8H4F15N
InChi Key: HZCZXRSVKNFILZ-UHFFFAOYSA-N
SMILES: NCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results