Learn More
1alpha, 25-Dihydroxy VD2-d6, MedChemExpress
Product Code. 16491499
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16491499
Brand: Medchem Express HY15327
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress 1alpha, 25-Dihydroxy VD2-D6 is a deuterated form of vitamin D.
1alpha, 25-Dihydroxy VD2-D6 is a deuterated form of vitamin D.
Specifications
Chemical Name or Material 1alpha, 25-Dihydroxy VD2-d6
CAS 216244-04-1
Purity Grade Notes Research
Molecular Formula C28H38D6O3
SMILES O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
Industry Type Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 434.68
Quantity 1 mg
Formula Weight 434.68
Grade Research