Learn More
17alpha-Boldenone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30323007
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323007 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323007 Supplier Dr. Ehrenstorfer Supplier No. DREA10662100AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 17alpha-Boldenone
CAS 27833-18-7
Analyte or Component Names 17alpha-Boldenone
Shipping Condition Room Temperature
Molecular Formula C19 H26 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym (17alpha)-17-Hydroxyandrosta-1,4-dien-3-one, 17alpha-Hydroxyandrosta-1,4-dien-3-one, 17alpha-Hydroxyandrosta-1,4-dien-3-one, Epiboldenone, alpha-Boldenone, EPIBOLDENONE
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@H]2O
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 286.4
Quantity 1 mL
Formula Weight 286.4085 g/mol