Learn More
Adrafinil, TRC
Product Code. 17953665
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953665 10 mg 10mg
30009217 25 mg 25mg
17930515 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17953665 Supplier Toronto Research Chemicals Supplier No. TRCA30150010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63547-13-7
Molecular Formula C15H15NO3S
Molecular Weight (g/mol) 289.35
Synonym Acetamide, 2-[(diphenylmethyl)sulfinyl]-N-hydroxy- (9CI, ACI), 2-[(Diphenylmethyl)sulfinyl]-N-hydroxyacetamide (ACI), 2-Benzhydrylsulfinyl-N-hydroxyacetamide, Adrafinil, CRL 40028, Olmifon
IUPAC Name 2-benzhydrylsulfinyl-N-hydroxyacetamide
SMILES ONC(=O)CS(=O)C(c1ccccc1)c2ccccc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C15H15NO3S/c17-14(16-18)11-20(19)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15,18H,11H2,(H,16,17)
Formula Weight 289.0773
Chemical Name or Material Adrafinil