CAS RN 63547-13-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Adrafinil, TRC
CAS: 63547-13-7 Molecular Formula: C15H15NO3S Molecular Weight (g/mol): 289.35 Synonym: Acetamide, 2-[(diphenylmethyl)sulfinyl]-N-hydroxy- (9CI, ACI),2-[(Diphenylmethyl)sulfinyl]-N-hydroxyacetamide (ACI),2-Benzhydrylsulfinyl-N-hydroxyacetamide,Adrafinil,CRL 40028,Olmifon IUPAC Name: 2-benzhydrylsulfinyl-N-hydroxyacetamide SMILES: ONC(=O)CS(=O)C(c1ccccc1)c2ccccc2