missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), Mikromol™

Product Code. 18090240
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090240 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090240 Supplier Mikromol Supplier No. MM3717.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone Cyclohexylmethylcarbonate
CAS 23454-33-3
Analyte or Component Names Trenbolone Cyclohexylmethylcarbonate
Shipping Condition Room Temperature
Molecular Formula C26 H34 O4
Certifications/Compliance ISO 17025
Synonym Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), Estra-4,9,11-trien-3-one, 17-[[(cyclohexylmethoxy)carbonyl]oxy]-, (17β)-, Estra-4,9,11-trien-3-one, 17β-hydroxy-, cyclohexylmethyl carbonate (8CI), Carbonic acid, cyclohexylmethyl ester, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Cyclohexanemethanol, hydrogen carbonate, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Parabolan, RU 2580, Trenbolone cyclohexylmethyl carbonate, Trenbolone hexahydrobenzyl carbonate
InChI Formula InChI=1S/C26H34O4/c1-26-14-13-21-20-10-8-19(27)15-18(20)7-9-22(21)23(26)11-12-24(26)30-25(28)29-16-17-5-3-2-4-6-17/h13-15,17,22-24H,2-12,16H2,1H3/t22-,23+,24+,26+/m1/s1
SMILES C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2OC(=O)OCC5CCCCC5
IUPAC Name cyclohexylmethyl [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] carbonate
Molecular Weight (g/mol) 410.55
Quantity 100 mg
Formula Weight 410.554 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.