missing translation for 'onlineSavingsMsg'
Learn More

ML355, MedChemExpress

Product Code. 16425836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425836

Brand: Medchem Express HY12341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML355 is a potent and selective inhibitor of 12-Lipoxygenase (12-LOX) with an IC50 of 0.34 μM, shows excellent selectivity over related lipoxygenases and cyclooxygenases, and possesses favorable ADME properties.

ML355 is a potent and selective inhibitor of 12-Lipoxygenase (12-LOX) with an IC50 of 0.34 μM, shows excellent selectivity over related lipoxygenases and cyclooxygenases, and possesses favorable ADME properties.

Specifications

Chemical Name or Material ML355
CAS 1532593-30-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H19N3O4S2
Quantity 100 mg
Solubility Information DMSO : ≥ 42 mg/mL (95.13 mM)
SMILES O=S(NC1=NC2=CC=CC=C2S1)(C3=CC=C(NCC4=CC=CC(OC)=C4O)C=C3)=O
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 95.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.