missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Ester of Cetirizine Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl Ester of Cetirizine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Methyl Ester of Cetirizine Dihydrochloride |
| CAS | 83881-48-5 |
| Analyte or Component Names | Methyl Ester of Cetirizine Dihydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H27 Cl N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl Ester of Cetirizine Dihydrochloride |
| InChI Formula | InChI=1S/C22H27ClN2O3.2ClH/c1-27-21(26)17-28-16-15-24-11-13-25(14-12-24)22(18-5-3-2-4-6-18)19-7-9-20(23)10-8-19;;/h2-10,22H,11-17H2,1H3;2*1H |
| SMILES | Cl.Cl.COC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | methyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction