missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibogaine Hydrochloride, Mikromol™
Description
Ibogaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Ibogaine Hydrochloride |
| CAS | 5934-55-4 |
| Analyte or Component Names | Ibogaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H26 N2 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ibogaine Hydrochloride, Ibogamine, 12-methoxy-, hydrochloride (1:1), Ibogaine, hydrochloride (7CI), Ibogaine, monohydrochloride (8CI), Ibogamine, 12-methoxy-, monohydrochloride (9CI), 6,9-Methano-5H-pyrido[1',2':1,2]azepino[4,5-b]indole, ibogamine deriv., NSC 29847 |
| InChI Formula | InChI=1S/C20H26N2O.ClH/c1-3-13-8-12-9-17-19-15(6-7-22(11-12)20(13)17)16-10-14(23-2)4-5-18(16)21-19;/h4-5,10,12-13,17,20-21H,3,6-9,11H2,1-2H3;1H/t12-,13+,17+,20+;/m1./s1 |
| SMILES | Cl.CC[C@H]1C[C@@H]2C[C@@H]3[C@H]1N(CCc4c3[nH]c5ccc(OC)cc45)C2 |
| IUPAC Name | (2R,4S,4aS,12bR)-4-ethyl-2,5-methano-9-methoxy-2,3,4,4a,6,7,12,12b-octahydro-1H-indolo[3,2-d][1]benzazepine monohydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?