missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexibuprofen Ethyl Ester, Mikromol™
Description
Dexibuprofen Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Dexibuprofen Ethyl Ester |
| CAS | 272458-63-6 |
| Analyte or Component Names | Dexibuprofen Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H22 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexibuprofen Ethyl Ester, (alphaS)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid ethyl ester, (S)-Ibuprofen ethyl ester |
| InChI Formula | InChI=1S/C15H22O2/c1-5-17-15(16)12(4)14-8-6-13(7-9-14)10-11(2)3/h6-9,11-12H,5,10H2,1-4H3/t12-/m0/s1 |
| SMILES | CCOC(=O)[C@@H](C)c1ccc(CC(C)C)cc1 |
| IUPAC Name | ethyl (2S)-2-[4-(2-methylpropyl)phenyl]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?