missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alvimopan Dihydrate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Alvimopan Dihydrate
Specifications
Specifications
| Chemical Name or Material | Alvimopan Dihydrate |
| CAS | 170098-38-1 |
| Analyte or Component Names | Alvimopan Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H32 N2 O4 . 2 H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Glycine, N-[(2S)-2-[[(3R,4R)-4-(3-hydroxyphenyl)-3,4-dimethyl-1-piperidinyl]methyl]-1-oxo-3-phenylpropyl]-, hydrate (1:2), Glycine, N-[(2S)-2-[[(3R,4R)-4-(3-hydroxyphenyl)-3,4-dimethyl-1-piperidinyl]methyl]-1-oxo-3-phenylpropyl]-, dihydrate (9CI), ADL 8-2698 dihydrate, Alvimopan dihydrate, LY 246736 dihydrate, Glycine, N-[2-[[4-(3-hydroxyphenyl)-3,4-dimethyl-1-piperidinyl]methyl]-1-oxo-3-phenylpropyl]-, dihydrate, [3R-[1(S*),3alpha,4alpha]]- |
| InChI Formula | InChI=1S/C25H32N2O4.2H2O/c1-18-16-27(12-11-25(18,2)21-9-6-10-22(28)14-21)17-20(24(31)26-15-23(29)30)13-19-7-4-3-5-8-19;;/h3-10,14,18,20,28H,11-13,15-17H2,1-2H3,(H,26,31)(H,29,30);2*1H2/t18-,20-,25+;;/m0../s1 |
| SMILES | O.O.C[C@H]1CN(C[C@H](Cc2ccccc2)C(=O)NCC(=O)O)CC[C@@]1(C)c3cccc(O)c3 |
| IUPAC Name | 2-[[(2S)-2-benzyl-3-[(3R,4R)-4-(3-hydroxyphenyl)-3,4-dimethylpiperidin-1-yl]propanoyl]amino]acetic acid;dihydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction