missing translation for 'onlineSavingsMsg'
Learn More

9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate), Mikromol™

Product Code. 17909919
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17909919

Brand: Mikromol MM0199.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beclometasone 17-Propionate
CAS 5534-18-9
Analyte or Component Names Beclometasone 17-Propionate
Shipping Condition Room Temperature
Molecular Formula C25 H33 Cl O6
Certifications/Compliance ISO 17025
Synonym 9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Beclometasone Dipropionate Imp. H (EP), Beclometasone 17-Propionate, Beclomethasone 17-monopropionate, Beclomethasone 17-propionate, (11beta,16beta)-9-Chloro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-propionate, CCI 2382, Beclometasone Dipropionate Related Compound H, Beclometasone Dipropionate Impurity H
InChI Formula InChI=1S/C25H33ClO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)CO
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 464.98
Quantity 100 mg
Formula Weight 464.1966 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.