missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amlodipine EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Amlodipine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 3-ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[(2-[(2-(methylcarbamoyl)benzoyl)amino]ethoxy)methyl]-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 721958-72-1 |
| Analyte or Component Names | 3-ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[(2-[(2-(methylcarbamoyl)benzoyl)amino]ethoxy)methyl]-1,4-dihydropyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H32 Cl N3 O7 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-6-methyl-2-[[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. B (EP), Amlodipine Imp. B (EP), 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-6-methyl-2-[[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Related Compound B, Amlodipine Besilate Impurity B, Amlodipine Impurity B |
| InChI Formula | InChI=1S/C29H32ClN3O7/c1-5-40-29(37)25-22(16-39-15-14-32-27(35)19-11-7-6-10-18(19)26(34)31-3)33-17(2)23(28(36)38-4)24(25)20-12-8-9-13-21(20)30/h6-13,24,33H,5,14-16H2,1-4H3,(H,31,34)(H,32,35) |
| SMILES | CCOC(=O)C1=C(COCCNC(=O)c2ccccc2C(=O)NC)NC(=C(C1c3ccccc3Cl)C(=O)OC)C |
| IUPAC Name | 5-O-ethyl 3-O-methyl 4-(2-chlorophenyl)-2-methyl-6-[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxymethyl]-1,4-dihydropyridine-3,5-dicarboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction