missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound), Mikromol™
Description
3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound)
Specifications
Specifications
| Chemical Name or Material | 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound) |
| CAS | 85045-98-3 |
| Analyte or Component Names | 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H23 Br N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Celiprolol Hydrochloride Imp. H (EP), Celiprolol Imp. H (EP), 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Bromhydrin Compound of Celiprolol, Celiprolol Hydrochloride Impurity H, Celiprolol Impurity H |
| InChI Formula | InChI=1S/C16H23BrN2O4/c1-4-19(5-2)16(22)18-12-6-7-15(14(8-12)11(3)20)23-10-13(21)9-17/h6-8,13,21H,4-5,9-10H2,1-3H3,(H,18,22) |
| SMILES | CCN(CC)C(=O)Nc1ccc(OCC(O)CBr)c(c1)C(=O)C |
| IUPAC Name | 3-[3-acetyl-4-(3-bromo-2-hydroxypropoxy)phenyl]-1,1-diethylurea |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?