missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-epi-Estradiol Valerate (17alpha-Estradiol Valerate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
17-epi-Estradiol Valerate (17alpha-Estradiol Valerate)
Specifications
Specifications
| Chemical Name or Material | 17-epi-Estradiol Valerate |
| CAS | 182624-54-0 |
| Analyte or Component Names | 17-epi-Estradiol Valerate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H32 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 17-epi-Estradiol Valerate, 17alpha-Estradiol Valerate |
| InChI Formula | InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21-,23+/m1/s1 |
| SMILES | CCCCC(=O)O[C@@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,13S,14S,17R)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction