missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketoprofen EP Impurity A (Ketoprofen USP Related Compound D), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ketoprofen EP Impurity A (Ketoprofen USP Related Compound D)
Specifications
Specifications
| Chemical Name or Material | 3-Acetylbenzophenone |
| CAS | 66067-44-5 |
| Analyte or Component Names | 3-Acetylbenzophenone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-(3-Benzoylphenyl)ethanone (3-Acetylbenzophenone), Ketoprofen Imp. A (EP), Ketoprofen USP Related Compound D, Ketoprofen USP RC D, 1-(3-Benzoylphenyl)ethanone, 3-Acetylbenzophenone, Ketoprofen Related Compound D, Ketoprofen Impurity A |
| InChI Formula | InChI=1S/C15H12O2/c1-11(16)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-10H,1H3 |
| SMILES | CC(=O)c1cccc(c1)C(=O)c2ccccc2 |
| IUPAC Name | 1-(3-benzoylphenyl)ethanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction