missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Mikromol™
Description
1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate
Specifications
Specifications
| Chemical Name or Material | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| CAS | 203264-09-9 |
| Analyte or Component Names | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H16 Cl2 N2 O . H N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-[(2RS)-2-Benzyloxy-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. H (EP) as Nitrate, Miconazole Impurity H as Nitrate, Miconazole Nitrate Impurity H as Nitrate, Miconazole Impurity H as Nitrate |
| InChI Formula | InChI=1S/C18H16Cl2N2O.HNO3/c19-15-6-7-16(17(20)10-15)18(11-22-9-8-21-13-22)23-12-14-4-2-1-3-5-14;2-1(3)4/h1-10,13,18H,11-12H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccccc3)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-phenylmethoxyethyl]imidazole;nitric acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?