Learn More
1-Methylethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Product Code. 17940906
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17940906 25 mg 25mg
17900616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17940906 Supplier Mikromol Supplier No. MM0084.110025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methylethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Methylethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
CAS 76508-35-5
Analyte or Component Names 1-Methylethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
Shipping Condition Room Temperature
Molecular Formula C12 H13 N O5 S
Certifications/Compliance ISO 17025
Synonym 1-Methylethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Piroxicam Imp. I (EP), Piroxicam Impurity I
InChI Formula InChI=1S/C12H13NO5S/c1-7(2)18-12(15)10-11(14)8-5-3-4-6-9(8)19(16,17)13-10/h3-7,13-14H,1-2H3
SMILES CC(C)OC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1
IUPAC Name propan-2-yl 4-hydroxy-1,1-dioxo-2H-1λ^{6},2-benzothiazine-3-carboxylate
Molecular Weight (g/mol) 283.3
Quantity 25 mg
Formula Weight 283.051 g/mol