Learn More
1-Methyl-2-(methylsulphanyl)-1H-imidazole, Mikromol™
Description
1-Methyl-2-(methylsulphanyl)-1H-imidazole
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-2-(methylsulfanyl)-1H-imidazole |
| CAS | 14486-52-3 |
| Analyte or Component Names | 1-Methyl-2-(methylsulfanyl)-1H-imidazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H8 N2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Methyl-2-(methylsulphanyl)-1H-imidazole, Thiamazole Imp. C (EP), 1H-Imidazole, 1-methyl-2-(methylthio)- (9CI), Imidazole, 1-methyl-2-(methylthio)- (8CI), 1-Methyl-2-(methylthio)imidazole, 2-Methylthio-3-methylimidazole, Methimazole USP Related Compound C, Methimazole USP RC C, Methimazole Related Compound C, Thiamazole Impurity C |
| InChI Formula | InChI=1S/C5H8N2S/c1-7-4-3-6-5(7)8-2/h3-4H,1-2H3 |
| SMILES | CSc1nccn1C |
| IUPAC Name | 1-methyl-2-methylsulfanylimidazole |
| Show More |