missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene, 99+%, Thermo Scientific™

Product Code. 11481710
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481710 10 g 10g
11471710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481710 Supplier Thermo Scientific Alfa Aesar Supplier No. H27038.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63221-88-5
Molecular Formula C17H16O
Molecular Weight (g/mol) 236.31
MDL Number MFCD00807428
InChI Key YWDALJMLXXWXFP-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-methoxyphenyl ethynyl benzene, 1-4-ethylphenyl-2-4-methoxyphenyl acetylene, 4-4-ethylphenyl ethynyl anisole, 1-ethyl-4-2-4-methoxyphenyl ethynyl benzene, benzene,1-ethyl-4-2-4-methoxyphenyl ethynyl, 1-2-4-ethylphenyl ethynyl-4-methoxybenzene, 4-methoxy-4'-ethyltolane, acmc-209ne1
PubChem CID 3952083
IUPAC Name 1-ethyl-4-[2-(4-methoxyphenyl)ethynyl]benzene
SMILES CCC1=CC=C(C=C1)C#CC1=CC=C(OC)C=C1

Specifications

Melting Point 35°C (N)
Boiling Point 151°C to 153°C (1 mmHg)
Quantity 10 g
Formula Weight 236.31
Percent Purity ≥99%
Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.