accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COCH3

CAS RN 63221-88-5

IUPAC Name: 1-ethyl-4-[2-(4-methoxyphenyl)ethynyl]benzene
Synonyms: 1-ethyl-4-[2-(4-methoxyphenyl)ethynyl]benzene
Molecular Weight (g/mol): 236.31
Molecular Formula: C17H16O
InChi Key: YWDALJMLXXWXFP-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C#CC1=CC=C(OC)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results