Learn More
1-Deoxy-L-altronojirimycin Hydrochloride, TRC
Product Code. 30363762
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363762 1 mg 1mg
30362785 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30363762 Supplier Toronto Research Chemicals Supplier No. TRCD2317501MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 355138-93-1
Molecular Formula C6 H13 N O4 . Cl H
Molecular Weight (g/mol) 199.63
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2S,3S,4R,5S)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2S,3S,4R,5S)- (9CI), [2S-(2α,3β,4β,5α)]-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride, (2S,3S,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride, L-Deoxyaltronojirimycin hydrochloride, 1-Deoxy-L-altronojirimycin hydrochloride
IUPAC Name (2S,3S,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.OC[C@@H]1NC[C@H](O)[C@@H](O)[C@H]1O
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4-,5-,6+;/m0./s1
Formula Weight 199.0611
Chemical Name or Material 1-Deoxy-L-altronojirimycin, Hydrochloride