CAS RN 355138-93-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Deoxy-L-altronojirimycin Hydrochloride, TRC
CAS: 355138-93-1 Molecular Formula: C6 H13 N O4 . Cl H Molecular Weight (g/mol): 199.63 Synonym: 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2S,3S,4R,5S)-,3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2S,3S,4R,5S)- (9CI),[2S-(2α,3β,4β,5α)]-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride,(2S,3S,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride,L-Deoxyaltronojirimycin hydrochloride,1-Deoxy-L-altronojirimycin hydrochloride IUPAC Name: (2S,3S,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride SMILES: Cl.OC[C@@H]1NC[C@H](O)[C@@H](O)[C@H]1O