Learn More
1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride (Desfluoroenrofloxacin Hydrochloride), Mikromol™
Description
1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride (Desfluoroenrofloxacin Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | Desfluoroenrofloxacin Hydrochloride |
| CAS | 2732310-93-7 |
| Analyte or Component Names | Desfluoroenrofloxacin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 N3 O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Enrofloxacin Imp. C (EP), 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Enrofloxacin Imp. C (EP) as Hydrochloride, Desfluoroenrofloxacin Hydrochloride, Enrofloxacin Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C19H23N3O3.ClH/c1-2-20-7-9-21(10-8-20)14-5-6-15-17(11-14)22(13-3-4-13)12-16(18(15)23)19(24)25;/h5-6,11-13H,2-4,7-10H2,1H3,(H,24,25);1H |
| Solution Type | Neat |
| SMILES | Cl.CCN1CCN(CC1)c2ccc3C(=O)C(=CN(C4CC4)c3c2)C(=O)O |
| Show More |