Learn More
1-Cyclopropyl-6,8-difluoro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1-Cyclopropyl-6,8-difluoro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopropyl-6,8-difluoro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid |
| CAS | 151213-15-9 |
| Analyte or Component Names | 1-Cyclopropyl-6,8-difluoro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 F2 N3 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Moxifloxacin Hydrochloride Imp. A (EP), Moxifloxacin Imp. A (EP), Moxifloxacin USP Related Compound A, Moxifloxacin USP RC A, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,8-difluoro-1,4-dihydro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,8-difluoro-1,4-dihydro-7-(octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-4-oxo-, (4aS-cis)-, 6H-Pyrrolo[3,4-b]pyridine, 3-quinolinecarboxylic acid deriv., Moxifloxacin Related Compound A, Moxifloxacin Hydrochloride Impurity A, Moxifloxacin Impurity A |
| InChI Formula | InChI=1S/C20H21F2N3O3/c21-14-6-12-17(25(11-3-4-11)8-13(19(12)26)20(27)28)16(22)18(14)24-7-10-2-1-5-23-15(10)9-24/h6,8,10-11,15,23H,1-5,7,9H2,(H,27,28)/t10-,15+/m0/s1 |
| SMILES | OC(=O)C1=CN(C2CC2)c3c(F)c(N4C[C@@H]5CCCN[C@@H]5C4)c(F)cc3C1=O |
| IUPAC Name | 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6,8-difluoro-4-oxoquinoline-3-carboxylic acid |
| Show More |