Learn More
1-(Chloromethyl)-4-(phenylsulfanyl)benzene, Mikromol™
Description
1-(Chloromethyl)-4-(phenylsulfanyl)benzene
Specifications
Specifications
| Chemical Name or Material | 1-(Chloromethyl)-4-(phenylsulfanyl)benzene |
| CAS | 1208-87-3 |
| Analyte or Component Names | 1-(Chloromethyl)-4-(phenylsulfanyl)benzene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H11 Cl S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzene, 1-(chloromethyl)-4-(phenylthio)-, 4-Phenylthiobenzyl chloride, NSC 43056, Sulfide, alpha-chloro-p-tolyl phenyl (7CI,8CI) |
| InChI Formula | InChI=1S/C13H11ClS/c14-10-11-6-8-13(9-7-11)15-12-4-2-1-3-5-12/h1-9H,10H2 |
| SMILES | ClCc1ccc(Sc2ccccc2)cc1 |
| IUPAC Name | 1-(chloromethyl)-4-phenylsulfanylbenzene |
| Show More |