Learn More
1-Bromo-4-n-propylbenzene, 99%
Product Code. 11346689
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346689 100 g 100g
11326689 5 g 5g
11336689 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11346689 Supplier Thermo Scientific Alfa Aesar Supplier No. H26236.22
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 588-93-2 | C9H11Br | 199.091 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 588-93-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00012456
InChI Key NUPWGLKBGVNSJX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
PubChem CID 136374
IUPAC Name 1-bromo-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)Br
Specifications
Density 1.286
Boiling Point 225°C
Flash Point 95°C (203°F)
Refractive Index 1.537
Quantity 100 g
Formula Weight 199.09
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-n-propylbenzene