CAS RN 588-93-2
CAS RN 588-93-2
IUPAC Name:
1-bromo-4-propylbenzene
Synonyms:
1-bromo-4-propylbenzene
Molecular Weight (g/mol):
199.09
Molecular Formula:
C9H11Br
InChi Key:
NUPWGLKBGVNSJX-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(Br)C=C1
1
–
2
of
2
results
1
1-Bromo-4-n-propylbenzene, 99%
CAS: 588-93-2 Molecular Formula: C9H11Br Molecular Weight (g/mol): 199.091 MDL Number: MFCD00012456 InChI Key: NUPWGLKBGVNSJX-UHFFFAOYSA-N Synonym: 1-bromo-4-n-propylbenzene,4-bromo-n-propylbenzene,1-4'-bromophenyl propane,1-bromo-4-propyl-benzene,benzene, 1-bromo-4-propyl,4-propylbromobenzene,4-propyl bromobenzene,4-n-propylbromobenzene,4-propylphenyl bromide,pubchem10762 PubChem CID: 136374 IUPAC Name: 1-bromo-4-propylbenzene SMILES: CCCC1=CC=C(C=C1)Br