missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-propylbenzene, 99%

Product Code. 11336689
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11336689

Brand: Thermo Scientific Alfa Aesar H26236.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 588-93-2 | C9H11Br | 199.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-93-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00012456
InChI Key NUPWGLKBGVNSJX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-propylbenzene, 4-bromo-n-propylbenzene, 1-4'-bromophenyl propane, 1-bromo-4-propyl-benzene, benzene, 1-bromo-4-propyl, 4-propylbromobenzene, 4-propyl bromobenzene, 4-n-propylbromobenzene, 4-propylphenyl bromide, pubchem10762
PubChem CID 136374
IUPAC Name 1-bromo-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)Br

Specifications

Density 1.286
Boiling Point 225°C
Flash Point 95°C (203°F)
Refractive Index 1.537
Quantity 25 g
Formula Weight 199.09
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-n-propylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.