missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(aminomethyl)pyrrolidine, 95%, Thermo Scientific™

Product Code. p-7030428
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433827 250 mg 250mg
11402110 1 g 1g
11412110 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433827 Supplier Thermo Scientific Alfa Aesar Supplier No. H28674.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270912-72-6
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD01861798
InChI Key OGCCBDIYOAFOGK-UHFFFAOYSA-N
Synonym tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate, 1-boc-3-aminomethyl pyrrolidine, 3-aminomethyl-1-n-boc-pyrrolidine, 1-boc-3-aminomrthylpyrrolidine, 1-boc-3-aminomethyl-pyrrolidine, +/--3-aminomethyl-1-n-boc-pyrrolidine, 1-n-boc-3-aminomethyl pyrrolidine, 1-boc-3-aminomethylpyrrolidine, 3-aminomethyl-1-boc-pyrrolidine, 3-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 2756485
IUPAC Name tert-butyl 3-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)CN

Specifications

Refractive Index 1.474
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material 1-Boc-3-(aminomethyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.