accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 270912-72-6

IUPAC Name: [(3R)-1-[(tert-butoxy)carbonyl]pyrrolidin-3-yl]methanaminium
Synonyms: [(3R)-1-(tert-butoxycarbonyl)pyrrolidin-3-yl]methanaminium
Molecular Weight (g/mol): 201.29
Molecular Formula: C10H21N2O2
InChi Key: OGCCBDIYOAFOGK-MRVPVSSYSA-O
SMILES: CC(C)(C)OC(=O)N1CC[C@H](C[NH3+])C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results