Learn More
1-Boc-3-(aminomethyl)azetidine, 97%, Thermo Scientific™
Product Code. 11422250
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422250 1 g 1g
15413847 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11422250 Supplier Thermo Scientific Alfa Aesar Supplier No. H29152.03
 Restricted Item
Estimated Shipment: 22-07-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 325775-44-8
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD01861762
InChI Key XSJPKMUFBHSIRA-UHFFFAOYSA-N
Synonym 1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
PubChem CID 10679125
IUPAC Name tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CN
Spezifikation
Refractive Index 1.467
Quantity 1 g
Formula Weight 186.25
Sensitivity Air Sensitive
Percent Purity 97%
Chemical Name or Material 1-Boc-3-(aminomethyl)azetidine