accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OONNH2

CAS RN 325775-44-8

IUPAC Name: tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: XSJPKMUFBHSIRA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(CN)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)