Learn More
1-Benzyl-4-phenylpiperidine-4-carboxylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1-Benzyl-4-phenylpiperidine-4-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1-Benzyl-4-phenylpiperidine-4-carboxylic Acid |
| CAS | 61886-17-7 |
| Analyte or Component Names | 1-Benzyl-4-phenylpiperidine-4-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Pethidine Hydrochloride Imp. F (EP), Pethidine Imp. F (EP), Isonipecotic acid, 1-benzyl-4-phenyl- (4CI), 1-Benzyl-4-phenyl-4-piperidinecarboxylic acid, 4-Phenyl-1-phenylmethylpiperidine-4-carboxylic acid, NSC 80690, 4-Piperidinecarboxylic acid, 4-phenyl-1-(phenylmethyl)-, Imp. F (EP): 1-Benzyl-4-phenylpiperidine-4-carboxylic Acid, Pethidine Hydrochloride Impurity F, Pethidine Impurity F |
| InChI Formula | InChI=1S/C19H21NO2/c21-18(22)19(17-9-5-2-6-10-17)11-13-20(14-12-19)15-16-7-3-1-4-8-16/h1-10H,11-15H2,(H,21,22) |
| SMILES | OC(=O)C1(CCN(Cc2ccccc2)CC1)c3ccccc3 |
| IUPAC Name | 1-benzyl-4-phenylpiperidine-4-carboxylic acid |
| Show More |