missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(4-hydroxyphenyl)piperazine, TRC

Product Code. 17983365
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983365 1 g 1g
30137592 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17983365 Supplier Toronto Research Chemicals Supplier No. TRCA1790451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67914-60-7
Formula Weight 220.1212
InChI Formula InChI=1S/C12H16N2O2/c1-10(15)13-6-8-14(9-7-13)11-2-4-12(16)5-3-11/h2-5,16H,6-9H2,1H3
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
Molecular Formula C12 H16 N2 O2
Molecular Weight (g/mol) 220.27
SMILES CC(=O)N1CCN(CC1)c2ccc(O)cc2
Synonym 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine

Specifications

Analyte or Component Names 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.