Learn More
1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]piperidine-4-carboxylic Acid Hydrochloride, Mikromol™
Description
1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]piperidine-4-carboxylic Acid Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]piperidine-4-carboxylic Acid Hydrochloride |
| CAS | 2724689-42-1 |
| Analyte or Component Names | 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]piperidine-4-carboxylic Acid Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H31 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Terfenadine Imp. E (EP), 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]piperidine-4-carboxylic Acid Hydrochloride, Terfenadine Imp. E (EP) as Hydrochloride, Terfenadine Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s3 |
| SMILES | Cl.CC(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(=O)O |
| IUPAC Name | 1-[4-(4-tert-butylphenyl)-4-hydroxybutyl]piperidine-4-carboxylic acid;hydrochloride |
| Show More |